Learn More
Cayman Chemical Methomyl-d3 10mg
SDP
Supplier:  Cayman Chemical 3957610 MG
Encompass_Preferred
An internal standard for the quantification of methomyl by GC- or LC-MS
Catalog No. 50-291-5929