Learn More
Medchemexpress LLC Methomyl-d3 | 1398109-07-3 | C5H7D3N2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY152048S5MG
Encompass_Preferred
Methomyl-d3 is a deuterium labeled compound used largely as a tracer for quantitation during the drug development process. Deuterium substitution has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-73690