Learn More
Alkali Scientific Methotrexate, 100 Milligrams
SDP
Supplier:  Alkali Scientific THO003CP
Encompass_Preferred
Methotrexate, 100mg, is a laboratory-grade reagent used in research applications to study its effects on cancer, autoimmune diseases, and cellular processes. It is a folic acid antagonist that inhibits the enzyme dihydrofolate reductase, thereby blocking DNA and RNA synthesis, which leads to cell death. Methotrexate is frequently used in cell culture studies, drug testing, and cancer research. The 100mg quantity is ideal for smaller-scale research, providing sufficient material for experiments investigating its cytotoxic effects, as well as its role in targeting rapidly dividing cells in tumor tissues.
Catalog No. 50-006-60872