Learn More
Selleck Chemical LLC Methotrexate S1210-500mg
SDP
Supplier:  Selleck Chemical LLC S1210500MG
Encompass_Preferred
Methotrexate analog of folic acid is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells It forms an inactive ternary complex with DHFR and NADPH Methotrexate (MTX) induces apoptosis
Catalog No. 50-310-8341