Learn More
Selleck Chemical LLC Methotrexate 10mM/1mL 59-05-2 NCI-C04671
SDP
Supplier:  Selleck Chemical LLC S121010MM/1ML
Encompass_Preferred
Methotrexate (MTX, NCI-C04671), analog of folic acid, is a nonspecific inhibitor of the dihydrofolate reductase(DHFR) of bacteria and cancerous cells as well as normal cells. It forms an inactive ternary complex with DHFR and NADPH. Methotrexate (MTX) induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0634286