Learn More
Cayman Chemical Methotrexate-d3 1mg
SDP
Supplier:  Cayman Chemical 223781 MG
Encompass_Preferred
An internal standard for the quantification of methotrexate by GC- or LC-MS
Catalog No. 50-296-4306