Learn More
Medchemexpress LLC Methotrexate-d3 (Amethopterin-d3) | 00-00-0 | >99.0% | C20H19D3N8O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14519S110MG
Encompass_Preferred
Methotrexate-d3 is a deuterium-labeled analogue of methotrexate used as a stable isotope tracer and internal standard for quantitative analysis by LC-MS, NMR, and related analytical methods. Supplied as a solid with high chemical and isotopic purity, it is intended for research and analytical applications.
  • Deuterium-labeled internal standard for quantitative LC-MS and NMR
  • High chemical and isotopic purity (>99.0%) for accurate quantitation
  • Solid form for stable storage and simple handling
  • Soluble in common analytical solvents such as DMSO
  • Suitable for tracer studies, method development, and validation
Catalog No. 50-002-59142