Learn More
Selleck Chemical LLC Methotrexate disodium 25mg 7413-34-5
SDP
Supplier:  Selleck Chemical LLC S509725MG
Encompass_Preferred
Methotrexate sodium, an inhibitor of tetrahydrofolate dehydrogenase, is an antineoplastic antimetabolite with immunosuppressant properties. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8396