Learn More
Apexbio Technology LLC Methoxamine hydrochloride 61-16-5 50mg
SDP
Supplier:  Apexbio Technology LLC A992250
Encompass_Preferred
Methoxamine hydrochloride (CAS 61-16-5) is a selective agonist of 1-adrenergic receptors leading to vasoconstriction by promoting contraction of vascular smooth muscle This action increases peripheral vascular resistance and raises arterial blood pressure Methoxamine hydrochloride is widely used in biomedical research to model and investigate mechanisms of vasopressor response cardiovascular regulation and adrenergic receptor pharmacology It is also applied experimentally to study hypotensive states particularly those associated with anesthetic or surgical procedures
Catalog No. 50-000-93973