Learn More
Akina Inc Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AK0021G
Encompass
mPEG-PLGA (Mw ~2000:4110 Da) LA:GA 65:35 Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC2827953