Learn More
Medchemexpress LLC Methoxy polyethylene glycol maleimide | 724722-89-8 | 3400 (average) Da | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW020780C10MG
Encompass_Preferred
Methoxy polyethylene glycol maleimide (average molecular weight 3400 Da) is a maleimide-terminated polyethylene glycol reagent for site-specific thiol conjugation. It reacts selectively with free thiols to form stable thioether bonds, enabling pegylation of proteins and peptides for bioconjugation and drug delivery research. Handle to minimize aqueous hydrolysis of the maleimide group.
Catalog No. 50-002-99484