Learn More
Medchemexpress LLC Methoxy polyethylene glycol succinimidyl carbonate | 1058691-00-1 | MFCD30828680 | 98.0% | 393.39 g/mol | C16H27NO10 | 50 MG

Supplier: Medchemexpress LLC HY13015050MG
m-PEG5-succinimidyl carbonate is a methoxy-terminated, 5-unit polyethylene glycol succinimidyl carbonate used as a short PEG linker and an NHS carbonate reagent for conjugation to primary amines. It provides a stable, non-cleavable spacer to improve solubility and reduce steric hindrance in bioconjugation workflows such as ADC and PROTAC synthesis.
- Non-cleavable 5-unit methoxy-PEG linker.
- Reactive succinimidyl carbonate for primary amine conjugation.
- Molecular weight 393.39 g/mol.
- Purity 98.0%.
- Appearance: colorless to light yellow liquid.
- Storage: pure form -20°C (stable 3 years); in solvent -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.