Learn More
Medchemexpress LLC Methoxy-x04 | 863918-78-9 | MFCD25563254 | 98.5% | 344.4 g/mol | C23H20O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10324025MG
Encompass_Preferred
Methoxy-X04 is a blood-brain barrier-permeant fluorescent probe that selectively binds beta-pleated sheets in dense core amyloid β (Aβ) plaques. It is used for in vivo imaging and for histological staining of Aβ plaques, tangles, and cerebrovascular amyloid, and exhibits high affinity for Aβ fibrils.
  • Blood-brain barrier permeant fluorescent probe.
  • Selectively binds beta-pleated sheets in amyloid β plaques.
  • Suitable for in vivo imaging and histological staining.
  • High binding affinity for Aβ fibrils (Ki = 26.8 nM).
  • High purity supply (>98% by HPLC).
Catalog No. 50-002-47877