Learn More
Medchemexpress LLC Methoxyacetic acid | 625-45-6 | MFCD00004308 | 100.0% | 90.08 g/mol | C3H6O3 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1009100G
Encompass_Preferred
Methoxyacetic acid is a metabolite of ethylene glycol monomethyl ether When the concentration of methoxyacetic acid reaches a certain level it can inhibit the respiratory function of hepatic mitochondria and testicular mitochondria Methoxyacetic acid is somewhat toxic[1][2]
Catalog No. 50-002-68129