Learn More
Medchemexpress LLC (Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt | 29684-56-8 | MFCD00077815 | 98.0% | 238.30 | C8H18N2O4S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW04008110G
Encompass_Preferred
Burgess reagent is a mild, selective dehydrating reagent used in organic synthesis to convert secondary and tertiary alcohols into alkenes under gentle conditions. It is widely used for substrates that are sensitive to strong acids or bases and is suitable for laboratory-scale synthesis and method development.
  • Mild, selective dehydration of secondary and tertiary alcohols to alkenes.
  • Operates under neutral to mildly basic conditions, minimizing side reactions.
  • Tolerant of many functional groups, reducing the need for protecting groups.
  • Solid reagent with good shelf stability for routine laboratory use.
  • Soluble in common organic solvents for straightforward reaction setup.
  • Suitable for intramolecular dehydrations and complex molecule synthesis.
Catalog No. 50-002-99790