Learn More
Medchemexpress LLC Methoxychlor-d6 | 106031-79-2 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1873S5MG
Encompass_Preferred
Methoxychlor-d6 is the deuterium labeled Methoxychlor, intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09905