Learn More
Medchemexpress LLC Methoxychlor-d6 | 106031-79-2 | MFCD12022229 | 98.2% | 351.69 g/mol | C16H9D6Cl3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1873S10MG
Encompass_Preferred
Methoxychlor-d6 is the deuterium-labeled isotopologue of Methoxychlor supplied as a solid research reagent for use as a stable isotope tracer or internal standard in analytical chemistry and mass spectrometry. It offers high chemical purity and isotopic labeling for accurate quantitation and metabolic studies.
  • Deuterium-labeled analog suitable as an internal standard.
  • High purity for accurate quantitation.
  • Soluble in ethanol (100 mg/mL); ultrasonic assistance may be needed.
  • Solid, white to off-white, convenient for small-scale handling.
  • Supplied in a 10 mg pack for analytical and research use.
Catalog No. 50-002-97021