Learn More
AdipoGen Methoxyfenozide, 161050-58-4, MFCD03792750, 100 mg
SDP
Supplier:  AdipoGen CDXM0133M100
Encompass_Preferred
MF: C22H28N2O3, MW: 368.5, ≥98%, Appearance: White powder. Diacylhydrazine insecticide. Ecdysone agonist that mimics the biological function of the natural insect molting hormone 20-hydroxyecdysone, causing premature molting and interfering with the normal growth and development in insects. Ecdysteroid agonist are also used as ligands for gene expression and gene therapy research. Compound can be used as analytical reference material.
Catalog No. 50-101-6222