Learn More
Medchemexpress LLC Methoxyfenozide | 161050-58-4 | 98.8% | 368.5 g/mol | C22H28N2O3 | 1 ML

Supplier: Medchemexpress LLC HY11738610MM 1ML
Methoxyfenozide is a non-steroidal ecdysone receptor agonist insecticide supplied as a ready-to-use 10 mM solution in DMSO for laboratory research applications. It interferes with insect growth and development and is used in studies of insecticidal activity and insect physiology.
- Ready-to-use 10 mM solution in DMSO for consistent dosing.
- Convenient 1 mL vial volume for small-scale assays.
- High purity, ≥98.8%, suitable for research use.
- Chemical formula C22H28N2O3; molecular weight 368.5 g/mol.
- Useful for studies of insect growth, development, and ovicidal activity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.