Learn More
Medchemexpress LLC Methoxypolyethylene glycol maleimide | 724722-89-8 | 3,400 Da (average) | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW020780C
Encompass_Preferred
Methoxypolyethylene glycol maleimide (average MW 3,400 Da) is a methoxy-terminated polyethylene glycol functionalized with a maleimide group for selective thiol conjugation. It is used for thiol-specific PEGylation of proteins and peptides in bioconjugation and drug delivery research. Because the maleimide can hydrolyze in aqueous media, reaction conditions should be optimized to preserve reactivity.
  • Selective thiol-reactive maleimide for site-specific conjugation.
  • Average molecular weight 3,400 Da for moderate steric shielding.
  • Methoxy-terminated monofunctional PEG prevents crosslinking.
  • Improves solubility and circulation time of conjugated biomolecules.
  • Compatible with standard bioconjugation buffers; optimize pH to limit hydrolysis.
  • Supplied as a solid for convenient storage and handling.
Catalog No. 50-003-03919