Learn More
Medchemexpress LLC Methyclothiazide | 135-07-9 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB05625MG
Encompass_Preferred
Methyclothiazide is an orally active antihypertensive and diuretic agent. It reduces the vascular response to endogenous vasoconstricting stimuli and acts against voltage-dependent Ca-channel (VDCC) activity in vitro.
  • Orally active antihypertensive agent
  • Diuretic agent
  • Reduces vascular response to endogenous vasoconstricting stimuli
  • Acts against voltage-dependent Ca-channel (VDCC) activity in vitro
  • Induces endothelium-dependent inhibition of vasoconstrictor responses
  • Shows inhibitory effect on contractile response
  • Affects vascular responses to extracellular Ca2+ under high-K⁺ depolarizing conditions
  • Reduces Ca2+ contracture
Catalog No. 50-004-07904