Learn More
Medchemexpress LLC Methyclothiazide | 135-07-9 | MFCD00242610 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB056210MM/1ML
Encompass_Preferred
Methyclothiazide is an orally active antihypertensive and diuretic agent. It functions by reducing the vascular response to endogenous vasoconstricting stimuli such as Norepinephrine. Additionally, Methyclothiazide is active against voltage-dependent Ca-channel (VDCC) in vitro.
  • Orally active antihypertensive agent
  • Diuretic agent
  • Reduces vascular response to vasoconstricting stimuli
  • Active against voltage-dependent Ca-channel (VDCC) in vitro
Catalog No. 50-004-09460