Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 1-acetyl-2-oxo-2,3-dihydro-1H-indole-6-carboxylate | 676326-36-6 | MFCD18251630 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY7569710MG

Encompass_Preferred

Nintedanib impurity 5 is a small-molecule impurity of the kinase inhibitor nintedanib supplied for laboratory research and analytical use only. It is intended as a reference or intermediate for synthetic and analytical workflows; not for human or clinical use. Store under the conditions specified in the certificate of analysis.

  • Impurity reference for analytical method development.
  • Intended for laboratory research use only; not for human use.
  • CAS number 676326-36-6.
  • Molecular formula C12H11NO4.
  • Molecular weight 233.22 g/mol.
  • Refer to the certificate of analysis for purity, storage, and pack size details.

Catalog No. 50-002-98668


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.