Learn More
Medchemexpress LLC Methyl 10-undecenoate | 111-81-9 | MFCD00016689 | 99.0% | 198.30 g/mol | C12H22O2 | 25 G

Supplier: Medchemexpress LLC 25GHYW068214
Methyl 10-undecenoate is a methyl ester of undec-10-enoic acid supplied as a high-purity reagent for research and organic synthesis. It is commonly used as a fragrance and flavor intermediate, and as a synthetic building block in laboratory-scale reactions.
- High purity: 99.0%.
- CAS number 111-81-9.
- Molecular formula C12H22O2; molecular weight 198.30 g/mol.
- Available in 10 g, 25 g, 50 g, and 100 g quantities.
- Used in flavor and fragrance synthesis and organic chemistry research.
- Suitable for laboratory and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.