Learn More
Medchemexpress LLC Methyl 2-(1H-indol-3-yl)acetate | 1912-33-0 | MFCD00022749 | 99.9% | 189.21 g·mol⁻¹ | C11H11NO2 | 1 ML

Supplier: Medchemexpress LLC HYW01522410MM/1ML
Methyl 2-(1H-indol-3-yl)acetate is an endogenous indole-derived metabolite provided for research use. It is supplied both as a 10 mM solution in DMSO and in solid reagent quantities, with manufacturer-stated purity and recommended storage conditions for stable handling in biochemical and metabolic studies.
- Endogenous indole-derived metabolite suitable for metabolic research.
- Available as 10 mM in DMSO and as solids (100 mg-10 g).
- High stated purity of 99.9%.
- Molecular formula C11H11NO2 and molecular weight 189.21 g·mol⁻¹.
- Storage recommendations provided for both pure form and in solvent.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.