Learn More
Medchemexpress LLC Methyl 2-(1H-indol-3-yl)acetate-d5 | 102415-39-4 | 99.1% | 194.24 | C11H6D5NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW015224S10MG
Encompass_Preferred
Methyl 2-(1H-indol-3-yl)acetate-d5 is the deuterium-labeled isotopologue of methyl 2-(1H-indol-3-yl)acetate, provided as a high-purity stable isotope standard for analytical chemistry. It is intended for use as an internal standard in mass spectrometry, tracer experiments, and quantitative metabolic studies.
  • High purity suitable for analytical applications.
  • Deuterium-labeled for isotope-dilution mass spectrometry.
  • Available in small quantities for standards and method development.
  • Stable when stored under recommended conditions.
  • Molecular weight 194.24 and formula C11H6D5NO2.
Catalog No. 50-003-02557