Learn More
Medchemexpress LLC Methyl 2-(3,4-dimethoxyphenyl)acetate | 15964-79-1 | MFCD00017206 | 98.3% | 210.23 g·mol-1 | C11H14O4 | 5 G

Supplier: Medchemexpress LLC HYW0420395G
Methyl homoveratrate (methyl 2-(3,4-dimethoxyphenyl)acetate) is a research-grade small molecule supplied as a solid-liquid mixture and used as a metabolite standard of RWJ-26240 for analytical and pharmacokinetic studies. It is provided at high purity for laboratory research and as a solution in DMSO for analytical applications.
- metabolite of RWJ-26240 detectable in plasma, urine, and faecal extracts.
- high reported purity (98.3%).
- molecular formula C11H14O4 and molecular weight 210.23 g·mol-1.
- available in research pack sizes, including 5 G and 1 mL (10 mM in DMSO).
- documentation available: COA and SDS for handling and analytical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.