Learn More
Medchemexpress LLC Methyl 2,3-O-isopropylidene-β-D-ribofuranoside | 4099-85-8 | 96.0% | 204.22 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1139495G
Encompass_Preferred
Methyl 2,3-O-Isopropylidene-β-D-ribofuranoside is a furanose derivative that serves as an important intermediate for the synthesis of compounds such as arsenic-containing ribosides. This high-purity chemical is suitable for various research applications.
  • Furanose derivative
  • Intermediate for synthesizing arsenic-containing ribosides
  • 96.0% purity
  • Soluble in DMSO (100 mg/mL)
  • Multiple in vivo dissolution protocols provided
  • Extensive documentation available
  • Cited in scientific publications
  • For research use only
Catalog No. 50-004-35642