Learn More
Medchemexpress LLC Methyl 2,3-O-isopropylidene-α-L-rhamnopyranoside | 14133-63-2 | MFCD08703844 | ≥98.0% | 218.25 g·mol⁻¹ | C10H18O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW24131525MG
Encompass_Preferred
Methyl 2,3-O-isopropylidene-α-L-rhamnopyranoside is a protected methyl rhamnopyranoside used as a carbohydrate building block in glycochemistry and glycobiology research. The 2,3-hydroxyls are masked as an isopropylidene acetal, enabling selective transformations at other positions and facilitating oligosaccharide assembly and derivatization. It is supplied as a small-scale reagent for laboratory synthesis and analytical work.
  • Protected 2,3-diol as isopropylidene acetal for selective chemistry.
  • Suitable building block for oligosaccharide synthesis and derivatization.
  • Offered as a stable, easy-to-handle solid for bench-top use.
  • Available in small research pack sizes for method development.
  • Typically supplied with lot-specific analytical data or certificate of analysis.
Catalog No. 50-002-80815