Learn More
Medchemexpress LLC Methyl 2,5-dihydroxycinnamate | 63177-57-1 | MFCD00132932 | 99.8% | 194.18 | C10H10O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10100610MG
Encompass_Preferred
Methyl 2,5-dihydroxycinnamate is a small-molecule research chemical described as an erbstatin analog that potently inhibits epidermal growth factor receptor (EGFR) kinase activity. It is supplied as a solid and as DMSO solutions for laboratory research and biochemical assays. Intended for research use only; not for human or clinical applications.
  • Potent inhibitor of EGFR kinase activity.
  • Erbstatin analog small-molecule structure.
  • Available as solid and as 10 mM solution in DMSO.
  • High purity suitable for analytical and biological assays.
  • Documented identifiers and physicochemical data for compound tracking.
Catalog No. 50-002-11854