Learn More
Medchemexpress LLC Methyl 2-acetamido-2-deoxy-alpha-D-glucopyranoside | 6082-04-8 | MFCD00056076 | >98.0% | 235.23 g/mol | C9H17NO6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1506931G
Encompass_Preferred
Methyl 2-acetamido-2-deoxy-alpha-D-glucopyranoside is a small-molecule biochemical reagent primarily used in glycobiology and synthetic organic chemistry. It functions as an N-acetylglucosaminide building block and analytical standard for research workflows, supplied as a purified solid suitable for laboratory handling and storage.
  • High purity (>98%) reagent suitable for analytical and synthetic applications.
  • Solid form with defined molecular weight for accurate dosing.
  • Useful as an N-acetylglucosaminide building block in glycochemistry.
  • Compatible with common organic solvents and standard storage conditions.
  • Available in multiple laboratory-scale package sizes for experimental flexibility.
Catalog No. 50-002-80579