Learn More
Medchemexpress LLC Methyl 2-aminoisobutyrate hydrochloride | 15028-41-8 | 153.61 | 25 G

Supplier: Medchemexpress LLC 25GHYW002520
Methyl 2-aminoisobutyrate hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Purity: 99.43%
- Molecular weight: 153.61
- Appearance: Solid, white to off-white
- Storage: -20°C, sealed storage, away from moisture; in solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
- Solubility (in vitro): DMSO: 100 mg/mL (651.00 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.