Learn More
Medchemexpress LLC Methyl 2-((diphenylmethylene)amino)acetate | 81167-39-7 | MFCD06661218 | 100.0% | 253.30 g/mol | C16H15NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW018045100G
Encompass_Preferred
Methyl 2-((diphenylmethylene)amino)acetate is a glycine-derived methyl ester used as a synthetic building block and intermediate in organic and peptide chemistry. Supplied as a high-purity reagent, it is employed in protecting-group strategies and the synthesis of substituted amino-acid derivatives; its molecular formula is C16H15NO2 and molecular weight is 253.30 g/mol.
  • High purity suitable for analytical and synthetic use.
  • Acts as a glycine-derived methyl ester building block.
  • Useful for protecting-group and amino-acid derivative synthesis.
  • Characterized by CAS 81167-39-7 and molecular formula C16H15NO2.
  • Available in gram-scale quantities for laboratory research.
Catalog No. 50-003-01399