Learn More
Medchemexpress LLC Methyl 2-(tert-butoxycarbonylamino)-3-carboxypropanoate | 98045-03-5 | MFCD03094777 | >97.0% | 247.25 | C10H17NO6 | 100 G

Supplier: Medchemexpress LLC HYW018741100G
Boc-Asp-OMe is N-Boc-L-aspartic acid 1-methyl ester, a Boc-protected aspartic acid methyl ester used as a building block in peptide and organic synthesis. Supplied as a white to off-white solid, it is intended for research use as a protected amino acid for selective coupling and deprotection strategies.
- Protected amino acid scaffold for selective deprotection reactions.
- Suitable for peptide synthesis and small-molecule synthesis workflows.
- Methyl ester functionality enables ester chemistry and selective transformations.
- Purity ≥97.0% suitable for research applications.
- Molecular weight 247.25 g/mol; formula C10H17NO6.
- White to off-white solid with recommended powder storage at -20°C (3 years) or 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.