Learn More
Medchemexpress LLC Methyl (2S)-2-[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]amino]propanoate | 778624-05-8 | >98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1206515MG
Encompass_Preferred
LDL-IN-2 is a small-molecule research compound reported to act as an antioxidant that prevents copper-mediated low-density lipoprotein (LDL) oxidation. It is supplied for research use only and is typically provided as a solid with reported purity greater than 98% and solubility in DMSO.
  • Antioxidant activity against copper-mediated LDL oxidation.
  • Reported purity greater than 98%.
  • Soluble in DMSO; suitable for common in vitro formulations.
  • Available in small research quantities such as 5 mg and 10 mg.
  • Intended for research use only, not for human or diagnostic use.
Catalog No. 50-002-30268