Learn More
Apexbio Technology LLC Methyl 3,4-dihydroxybenzoate 2150-43-8 50mg
SDP
Supplier:  Apexbio Technology LLC A478950
Encompass_Preferred
Methyl 3 4-dihydroxybenzoate (CAS 2150-43-8) also known as protocatechuic acid methyl ester is a hydroxybenzoate compound derived from the metabolic processing of tea polyphenols It exhibits antioxidative properties by directly scavenging free radicals and inhibiting oxidative stress-related enzymatic activity In addition it modulates inflammatory signaling pathways contributing to anti-inflammatory effects This molecule is widely employed in cellular and animal models to investigate oxidative injury elucidate mechanisms underlying inflammation and explore therapeutic strategies for related diseases
Catalog No. 50-000-93515