Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 3-methoxyacrylate | 34846-90-7 | 116.12 | 1 KG
SDP

Supplier:  Medchemexpress LLC HYW0079721KG

Encompass_Preferred

Methyl 3-methoxyacrylate (3-Methoxyacrylic Acid Methyl Ester) is a Lignans product that can be isolated from the herbs of *Lysimachia clethroides*. This chemical is a valuable compound for various laboratory applications.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage: Room temperature for 3 years; in solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-48622


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.