Learn More
Medchemexpress LLC Methyl 3-O-methylgallate | 3934-86-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6669100MG
Encompass_Preferred
Methyl 3-O-methylgallate (M3OMG) is an analytical standard. This product is intended for research and analytical applications. Methyl 3-O-methylgallate possesses antioxidant effect and can protect neuronal cells from oxidative damage.
  • Intended for research and analytical applications.
  • Possesses antioxidant effect.
  • Can protect neuronal cells from oxidative damage.
  • The compound is a reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-18907