Learn More
Medchemexpress LLC Methyl 3-O-methylgallate | 3934-86-9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN666950MG
Encompass_Preferred
Methyl 3-O-methylgallate (M3OMG) is an analytical standard intended for research and analytical applications. It possesses antioxidant effects and can protect neuronal cells from oxidative damage.
  • It is the analytical standard of Methyl 3-O-methylgallate.
  • Intended for research and analytical applications.
  • Possesses antioxidant effect.
  • Can protect neuronal cells from oxidative damage.
  • It is used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-18820