Learn More
Medchemexpress LLC Methyl 4-(2-(3,4-dichlorophenyl)acetyl)-3-(1-pyrrolidinylmethyl)-1-piperazinecarboxylate fumarate | 126766-32-3 | MFCD00274044 | 99.9% | 530.4 | C23H29Cl2N3O7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10774710MM 1ML
Encompass_Preferred
GR 89696 (fumarate) is a potent, selective kappa-opioid receptor agonist used in preclinical pharmacology and pruritus research. It is provided as a fumarate salt and offered in solid form or as a ready-to-use 10 mM solution in DMSO, with solubility and storage data to support in vitro and in vivo studies.
  • Highly selective kappa-opioid receptor agonist with reported kappa2 preference.
  • Available as solid or ready-to-use 10 mM solution in DMSO.
  • High solubility in DMSO (≥ 50 mg/mL) for convenient stock preparation.
  • Provides storage and handling recommendations for stability.
  • Suitable for in vitro and in vivo pharmacology and pruritus models.
  • Supplied with standard chemical identifiers and characterization data.
Catalog No. 50-002-42404