Learn More
Accela Chembio Inc Methyl 4-bromo-2-methylbenzoate | 5g | 99548-55-7 | MFCD09954957 | 97+% | Shelf Life: 1260 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0080125G
Encompass_Preferred
Methyl 4-bromo-2-methylbenzoate | 5g | 99548-55-7 | MFCD09954957 | 97+% | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50-251-387