Learn More
Medchemexpress LLC Methyl 4-formylbenzoate | 1571-08-0 | 99.99% | C9H8O3 | 500 G
SDP
Supplier:  Medchemexpress LLC HY78000500G
Encompass_Preferred
Methyl 4-formylbenzoate (also known as 4-Carbomethoxybenzaldehyde) is an intermediate chemical used in research. It serves as a precursor for the synthesis of chalcone amide α-glucosidase inhibitors.
  • Used in scientific research.
  • An intermediate chemical for further synthesis.
  • It has a molecular weight of 164.16 and a chemical formula of C9H8O3.
  • Available as a white to off-white solid.
Catalog No. 50-004-34845