Learn More
Medchemexpress LLC Methyl 4-hydroxybenzoate | 99-76-3 | 152.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN03495G
Encompass_Preferred
Methyl paraben is a standardized allergen isolated from Yunnan hemlock (Tsuga dumosa) and commonly used as a stable, non-volatile preservative. It has been shown to increase histamine release and cellular regulation of immunity, block sodium channels, and prevent ischemia-reperfusion injury.
  • Standardized allergen isolated from Yunnan hemlock.
  • Functions as a stable, non-volatile preservative.
  • Increases histamine release and cellular regulation of immunity.
  • Blocks sodium channels.
  • Prevents ischemia-reperfusion injury.
Catalog No. 50-003-90077