Learn More
Medchemexpress LLC Methyl 4-hydroxybenzoate-d4 | 362049-51-2 | MFCD01861573 | 98.0% | 10 MG

Supplier: Medchemexpress LLC HYN0349S10MG
Methyl paraben-d4 (methyl 4-hydroxybenzoate-d4) is the deuterium-labeled analogue of methyl paraben, supplied as a small solid for use as an analytical/internal standard in chromatography and mass spectrometry. It provides a stable isotope reference for quantitative analyses and method development in laboratory workflows.
- Deuterium-labeled analogue (d4) for use as an internal standard.
- Molecular formula: C8H4D4O3.
- Molecular weight: 156.17 g/mol.
- CAS number: 362049-51-2.
- Supplied as a 10 mg vial suitable for analytical applications.
- Reported isotopic enrichment: ~98 atom % D (supplier listings).
- Intended applications include LC-MS and GC-MS quantitation and method validation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.