Learn More
Medchemexpress LLC Methyl 4-hydroxyphenylacetate | 14199-15-6 | MFCD00002387 | 166.18 | 250 MG
Supplier: Medchemexpress LLC 250MGHYW001084R
Methyl 4-hydroxyphenylacetate is a natural compound, a methyl ester resulting from the formal condensation of the carboxy group of 4-Hydroxyphenylacetic acid with methanol. It is for research use only.
- Colorless to light yellow liquid appearance
- High purity of 99.87% (HPLC)
- Derived from microorganisms, specifically mangrove-derived fungus Eupenicillium sp. HJ002
- Classified as a phenol and monophenol