Learn More
TARGETMOL CHEMICALS INC Methyl 5-hydroxy-1H-indole-3-c
SDP
Supplier:  TARGETMOL CHEMICALS INC TN71381MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Methyl 5-hydroxy-1H-indole-3-carboxylate (5-hydroxy-1H-indole-3-carboxylic acid methyl ester) is a marine derived natural products found in Hyrtios erectus. Purity 90%
Catalog No. 50-287-9397