Learn More
Medchemexpress LLC Methyl 5-hydroxypyridine-2-carboxylate | 30766-12-2 | 153.14 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0059635G
Encompass_Preferred
Methyl 5-hydroxypyridine-2-carboxylate is a phenolic acid found in the stems of Mahonia fortune. It exhibits NO inhibitory effects in vitro. This compound inhibits NO in LPS-stimulated RAW264.7 and BV2 cells, with IC50s of 115.67 and 118.80 μM, respectively. It does not exhibit any cytotoxic effect at concentrations of 6.25-200 μM.
  • Inhibits NO in LPS-stimulated RAW264.7 and BV2 cells
  • Does not show cytotoxic effect at concentrations of 6.25-200 μM
  • Found in the stems of Mahonia fortune
Catalog No. 50-004-46567