Learn More
Medchemexpress LLC Methyl 6-amino-6-deoxy-2,3-di-O-tetradecyl-alpha-D-glucopyranoside | 1115270-63-7 | MFCD31813901 | 98.0% | 585.94 g/mol | C35H71NO5 | 25MG

Supplier: Medchemexpress LLC HY12517125MG
IAXO-102 is a small-molecule TLR4 antagonist supplied for research use that inhibits TLR4-mediated signaling, reducing MAPK and NF-κB (p65) phosphorylation and downstream inflammatory protein expression in vitro and in vivo.
- High purity (98.0%) suitable for research applications.
- Off-white solid with molecular formula C35H71NO5 and molecular weight 585.94 g/mol.
- Soluble in ethanol and DMSO; ethanol 50 mg/mL and DMSO 3.33 mg/mL (may require ultrasonic or warming).
- Provides example in vivo vehicle formulations that achieve clear solutions at ≥0.83 mg/mL.
- Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years).
- Suitable for pharmacology and immunology studies of TLR4 signaling and inflammation models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.