Learn More
eMolecules​ METHYL 6-BROMO-2-FLUORO-3-METHOXYBENZOATE | AstaTech | 1007455-28-8 | MFCD20482377 | 263.062 | C9H8BrFO3 | 95.000 | COC(=O)c1c(Br)ccc(OC)c1F | 1g | 213338631
Supplier:  eMolecules​ 213338631
Encompass_Preferred
METHYL 6-BROMO-2-FLUORO-3-METHOXYBENZOATE | AstaTech | 1007455-28-8 | MFCD20482377 | 263.062 | C9H8BrFO3 | 95.000 | COC(=O)c1c(Br)ccc(OC)c1F | 1g | 213338631
Catalog No. NC1467026