Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyl 6-bromopicolinate | 26218-75-7 | 99.9% | C7H6BrNO2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHY76562

Encompass_Preferred

Methyl 6-bromopicolinate is a biochemical reagent suitable for various life science-related research applications. This organic compound serves as a biological material in scientific studies.

  • Can be used as a biochemical reagent
  • Appears as a white to off-white solid
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO, requiring ultrasonic treatment

Catalog No. 50-003-58680


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.